Merck
  • Transport-metabolism interplay: LXRalpha-mediated induction of human ABC transporter ABCC2 (cMOAT/MRP2) in HepG2 cells.
Transport-metabolism interplay: LXRalpha-mediated induction of human ABC transporter ABCC2 (cMOAT/MRP2) in HepG2 cells.
Molecular pharmaceutics (2009-07-11)
Tatsuhiko Adachi, Hiroshi Nakagawa, Yuichiro Hagiya, Tatsuya Yasuoka, Toshihisa Ishikawa
ABSTRACT
Human ATP-binding cassette (ABC) transporter ABCC2 (cMOAT/MRP2) plays a crucial role in the hepatobiliary transport of sulfate-, glucuronide-, and glutathione-conjugated metabolites as well as a variety of amphiphilic organic anions derived from hepatic metabolism. Molecular mechanisms underlying the induction of this hepatic ABC transporter are of great interest to understand the transport-metabolism interplay in vivo. In the present study, to gain insight into the mechanism of ABCC2 induction, we tested a total of 46 structurally diverse compounds, including nuclear receptor ligands, antibiotics, bile salts, phytochemicals, and anticancer drugs. Among them, we found that LXRalpha ligands, i.e., T0901317, paxilline, and 22(R)-hydroxycholesterol, acted potently to induce the expression of ABCC2 at both mRNA and protein levels in human hepatocellular carcinoma HepG2 cells. The ABCC2 induction by T0901317 was dose- and time-dependent, where the induction pattern of ABCC2 was very similar to that of ABCG1, one of the target genes of LXRalpha. The ABCC2 induction by T0901317 was more strongly elicited when the LXRalpha gene was transiently transfected into HepG2 cells. In contrast, ABCC2 induction by T0901317 was attenuated by transient transfection of a dominant negative LXRalpha variant, suggesting that LXRalpha is involved in ABCC2 induction. Interestingly, RXR, a heterodimer partner of LXRalpha, affected the mRNA levels of ABCC2 and ABCG1 differently. ABCC2 induction by T0901317 was enhanced by RXR siRNA treatment, whereas ABCG1 induction was suppressed by the same treatment. This is the first report demonstrating that LXRalpha is potentially involved in ABCC2 induction.
Sigma-Aldrich
22(R)-Hydroxycholesterol, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
22(S)-Hydroxycholesterol
Pricing and availability is not currently available.