Merck
  • Solution NMR structural study of a mixed aggregate of N-lithium triphenylphosphazene and lithium bromide.
Solution NMR structural study of a mixed aggregate of N-lithium triphenylphosphazene and lithium bromide.
Dalton transactions (Cambridge, England : 2003) (2009-03-18)
Ignacio Fernández, Matthew G Davidson, Richard D Price, Fernando López Ortiz
ABSTRACT
The toluene solution structure of an N-lithio(triphenyl)phosphazene (Ph(3)PNLi) mixed aggregate with lithium bromide (LiBr), 5, has been elucidated for the first time based on multinuclear magnetic resonance measurements ((1)H, (6)Li, (7)Li, (13)C and (31)P). The structure consist of two dimers [Li(mu-Z)](2) (Z= Br, NPR(3)) linked through LiX (X= N, Br) bridges. This arrangement is a cubane-like structure analogous to that found in the solid-state.
Sigma-Aldrich
Lithium bromide solution, 54 wt. % in H2O
Sigma-Aldrich
Lithium bromide, powder and chunks, ≥99.995% trace metals basis
Sigma-Aldrich
Lithium bromide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Lithium bromide, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium bromide, ReagentPlus®, ≥99%