Merck
  • Films of agarose enable rapid formation of giant liposomes in solutions of physiologic ionic strength.
Films of agarose enable rapid formation of giant liposomes in solutions of physiologic ionic strength.
Journal of the American Chemical Society (2009-01-22)
Kim S Horger, Daniel J Estes, Ricardo Capone, Michael Mayer
ABSTRACT
This paper describes a method to form giant liposomes in solutions of physiologic ionic strength, such as phosphate buffered saline (PBS) or 150 mM KCl. Formation of these cell-sized liposomes proceeded from hybrid films of partially dried agarose and lipids. Hydrating the films of agarose and lipids in aqueous salt solutions resulted in swelling and partial dissolution of the hybrid films and in concomitant rapid formation of giant liposomes in high yield. This method did not require the presence of an electric field or specialized lipids; it generated giant liposomes from pure phosphatidylcholine lipids or from lipid mixtures that contained cholesterol or negatively charged lipids. Hybrid films of agarose and lipids even enabled the formation of giant liposomes in PBS from lipid compositions that are typically problematic for liposome formation, such as pure phosphatidylserine, pure phosphatidylglycerol, and asolectin. This paper discusses biophysical aspects of the formation of giant liposomes from hybrid films of agarose and lipids in comparison to established methods and shows that gentle hydration of hybrid films of agarose and lipids is a simple, rapid, and reproducible procedure to generate giant liposomes of various lipid compositions in solutions of physiologic ionic strength without the need for specialized equipment.
Sigma-Aldrich
1,2-Dioleoyl-sn-glycero-3-phospho-L-serine sodium salt, ~95%