Merck
  • Energetics of conformational conversion between 1,1,2-trichloroethane polymorphs.
Energetics of conformational conversion between 1,1,2-trichloroethane polymorphs.
Chemical communications (Cambridge, England) (2008-09-20)
Maciej Bujak, Marcin Podsiadło, Andrzej Katrusiak
ABSTRACT
Pressure-induced transformations between gauche-, gauche+ and transoid conformations have been evidenced by X-ray single-crystal diffraction for 1,1,2-trichloroethane, and the energies of intermolecular interactions, conformational conversion, and the latent heat have been determined.
Product Description
Supelco
1,1,2-Trichloroethane solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
1,1,2-Trichloroethane, contains ≤3% 2-propanol as stabilizer, 97%
Supelco
1,1,2-Trichloroethane, analytical standard