Merck
Investigation of DNA as a catalyst for Henry reaction in water.
Chemical communications (Cambridge, England) (2008-08-08)
Jinmin Fan, Gaojun Sun, Changfeng Wan, Zhiyong Wang, Yingfu Li
ABSTRACT
Double-stranded DNA of natural origin can be used to facilitate nitro-aldol (or Henry) reaction in aqueous solution.
Sigma-Aldrich
4-Nitrobenzaldehyde, 98% (GC)
Supelco
4-Nitrobenzaldehyde, for spectrophotometric det. of amino sugars, ≥99.0%