Merck
  • Conducting nanocomposites of poly(N-vinylcarbazole) with single-walled carbon nanotubes.
Conducting nanocomposites of poly(N-vinylcarbazole) with single-walled carbon nanotubes.
Journal of nanoscience and nanotechnology (2008-06-25)
Arjun Maity, Suprakas Sinha Ray
ABSTRACT
The in situ solid-state polymerization of N-vinylcarbazole (NVC) at an elevated temperature in the presence of single-walled carbon nanotubes (SWCNTs) leads to the formation of new types of composite materials, the morphology and properties of which were characterized by field-emission scanning electron microscopy (FE-SEM), transmission electron microscopy (TEM), Fourier transform infrared (FTIR) spectroscopy, X-ray photoelectron spectroscopy (XPS), thermogravimetric analysis (TGA), and electrical property measurements. FTIR spectroscopy and XPS studies confirmed the ability of SWCNTs to initiate the in situ polymerization of NVC monomers. FE-SEM and TEM results showed the coating of the outer surfaces of SWCNTs by the PNVC hompolymer with separation of individual SWCNTs from the bundles. Thermogravimetric analysis revealed a moderate improvement in the thermal stability of the nanocomposites at a higher temperature region relative to the base polymer. The electrical conductivity of neat polymer dramatically improved in the presence of SWCNTs. For example, dc electrical conductivity increased from 10(-16)-10(-12) S x cm(-1) for neat PNVC to approximately 10(-6) S x cm(-1) for nanocomposite containing 9 wt% SWCNTs.
Sigma-Aldrich
Poly(9-vinylcarbazole), average Mw ~1,100,000, powder
Sigma-Aldrich
Poly(9-vinylcarbazole), average Mn 25,000-50,000