Merck
  • Differential metabolomics using stable isotope labeling and two-dimensional gas chromatography with time-of-flight mass spectrometry.
Differential metabolomics using stable isotope labeling and two-dimensional gas chromatography with time-of-flight mass spectrometry.
Analytical chemistry (2007-12-07)
Xiaodong Huang, Fred E Regnier
ABSTRACT
This work describes an approach to differential metabolomics that involves stable isotope labeling for relative quantification as part of sample analysis by two-dimensional gas chromatography/mass spectrometry (GCxGC/MS). The polar metabolome in control and experimental samples was extracted and differentially derivatized using isotopically light and heavy (D6) forms of the silylation reagent N-methyl-N-tert-butyldimethylsilyl)trifluoroacetamide (MTBSTFA). MTBSTFA derivatives are of much greater hydrolytic stability than the more common trimethylsilyl derivatives, thus diminishing the possibility of isotopomer scrambling during GC analysis. Subsequent to derivatization with MTBSTFA, differentially labeled samples were mixed and analyzed by GCxGC/MS. Metabolites were identified, and the isotope ratio of isotopomers was quantified. The method was tested using three classes of metabolites; amino acids, fatty acids, and organic acids. The relative concentration of isotopically labeled metabolites was determined by isotope ratio analysis. The accuracy and precision, respectively, in quantification of standard mixtures was 9.5 and 4.77% for the 16 amino acids, 9.7 and 2.83% for the mixture of 19 fatty acids, and 14 and 4.53% for the 20 organic acids. Suitability of the method for the examination of complex samples was demonstrated in analyses of the spiked blood serum samples. This differential isotope coding method proved to be an effective means to compare the concentration of metabolites between two samples simultaneously.
Product Description
Supelco
MTBSTFA (with 1% t-BDMCS), ampule of 5 × 1 mL, (with 1% t-BDMCS), analytical standard, Cerilliant®
Sigma-Aldrich
N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide with 1% tert-Butyldimethylchlorosilane, ≥95%
Sigma-Aldrich
N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide, >97%