Merck
Fragrance material review on trans-beta-damascone.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2007-11-23)
A Lapczynski, J Lalko, D McGinty, S Bhatia, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of trans-beta-damascone when used as a fragrance ingredient is presented.
Sigma-Aldrich
Damascenone, natural, 1.1-1.4 wt. % (190 proof ethanol), FG