Merck
  • Lung-targeting delivery of dexamethasone acetate loaded solid lipid nanoparticles.
Lung-targeting delivery of dexamethasone acetate loaded solid lipid nanoparticles.
Archives of pharmacal research (2007-05-11)
Qing-yu Xiang, Min-ting Wang, Fu Chen, Tao Gong, Yan-lin Jian, Zhi-rong Zhang, Yuan Huang
ABSTRACT
The objective of the present study was to develop a novel solid lipid nanoparticle (SLN) for the lung-targeting delivery of dexamethasone acetate (DXM) by intravenous administration. DXM loaded SLN colloidal suspensions were prepared by the high pressure homogenization method. The mean particle size, drug loading capacity and drug entrapment efficiency (EE%) of SLNs were investigated. In vitro drug release was also determined. The biodistribution and lung-targeting efficiency of DXM-SLNs and DXM-solutions (DXM-sol) in mice after intravenous administration were studied using reversed-phase high-performance liquid chromatography (HPLC). The results (expressed as mean +/- SD) showed that the DXM-SLNs had an average diameter of 552 +/- 6.5 nm with a drug loading capacity of 8.79 +/- 0.04% and an entrapment efficiency of 92.1 +/- 0.41%. The in vitro drug release profile showed that the initial burst release of DXM from DXM-SLNs was about 68% during the first 2 h, and then the remaining drug was released gradually over the following 48 hours. The biodistribution of DXM-SLNs in mice was significantly different from that of DXM-sol. The concentration of DXM in the lung reached a maximum level at 0.5 h post DXM-SLNs injection. A 17.8-fold larger area under the curve of DXM-SLNs was achieved compared to that of DXM-sol. These results indicate that SLN may be promising lung-targeting drug carrier for lipophilic drugs such as DXM.
Sigma-Aldrich
Dexamethasone 21-acetate, ≥99%
Dexamethasone acetate, European Pharmacopoeia (EP) Reference Standard