Merck
  • Chiral Diphenylprolinol TES ether promoted conjugate addition-aldol-dehydration reactions between alpha,beta-unsaturated aldehydes and 2-N-protected amino benzaldehydes.
Chiral Diphenylprolinol TES ether promoted conjugate addition-aldol-dehydration reactions between alpha,beta-unsaturated aldehydes and 2-N-protected amino benzaldehydes.
Organic letters (2007-02-15)
Hao Li, Jian Wang, Hexin Xie, Liansuo Zu, Wei Jiang, Eileen N Duesler, Wei Wang
ABSTRACT
A conjugate addition-aldol-dehydration reaction of alpha,beta-unsaturated aldehydes with 2-N-protected amino benzaldehydes has been developed. The process is promoted by (S)-diphenylprolinol TES ether to afford synthetically useful 1,2-dihydroquinolines in high enantioselectivities with good yields. [reaction: see text]
Sigma-Aldrich
(S)-(+)-2-Pyrrolidinemethanol, 97%
Sigma-Aldrich
TES, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
TES, BioPerformance Certified, suitable for cell culture, ≥99% (titration)
Sigma-Aldrich
TES, ≥99% (titration)