Merck
  • Selective monoiodination of aromatic compounds with electrochemically generated I+ using micromixing.
Selective monoiodination of aromatic compounds with electrochemically generated I+ using micromixing.
Chemical communications (Cambridge, England) (2006-09-14)
Koji Midorikawa, Seiji Suga, Jun-ichi Yoshida
ABSTRACT
Selective monoiodination of aromatic compounds such as dimethoxybenzene has been successfully achieved with I+, which is generated by anodic oxidation of I2 in acetonitrile, using micromixing.
Sigma-Aldrich
1,4-Dimethoxybenzene, 99%, FG
Sigma-Aldrich
1,4-Dimethoxybenzene, ReagentPlus®, 99%