Merck
  • Thermospray flame furnace-AAS determination of copper after on-line sorbent preconcentration using a system optimized by experimental designs.
Thermospray flame furnace-AAS determination of copper after on-line sorbent preconcentration using a system optimized by experimental designs.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2005-12-02)
César Ricardo Teixeira Tarley, Eduardo da Costa Figueiredo, Geraldo Domingues Matos
ABSTRACT
The present paper describes the on-line coupling of a flow-injection system to a new technique, thermospray flame furnace-AAS (TS-FF-AAS), for the preconcentration and determination of copper in water samples. Copper was preconcentrated onto polyurethane foam (PUF) complexed with ammonium O,O-diethyldithiophosphate (DDTP), while elution was performed using 80% (v/v) ethanol. An experimental design for optimizing the copper preconcentration system was established using a full factorial (2(4)) design without replicates for screening and a Doehlert design for optimization, studying four variables: sample pH, ammonium O,O-diethyldithiophosphate (DDTP) concentration, presence of a coil and the sampling flow rate. The results obtained from the full factorial and based on a Pareto chart indicate that only the pH and the DDTP concentration, as well as their interaction, exert influence on the system within a 95% confidence level. The proposed method provided a preconcentration factor of 65 fold, thus notably improving the detectability of TS-FF-AAS. The detection limit was 0.22 microg/dm3 and the precision, expressed as the relative standard deviation (RSD) for eight independent determinations, was 2.7 and 1.1 for copper solutions containing 5 and 30 microg/dm3, respectively. The procedure was successfully applied for copper determination in water samples.
Sigma-Aldrich
O,O′-Diethyl dithiophosphate, technical grade, 90%
Sigma-Aldrich
Diethyl dithiophosphate ammonium salt, 95%