Merck
  • Dihydrofolate reductase: x-ray structure of the binary complex with methotrexate.
Dihydrofolate reductase: x-ray structure of the binary complex with methotrexate.
Science (New York, N.Y.) (1977-07-29)
D A Matthews, R A Alden, J T Bolin, S T Freer, R Hamlin, N Xuong, J Kraut, M Poe, M Williams, K Hoogsteen
PMID17920
ABSTRACT
A central eight-stranded beta-pleated sheet is the main feature of the polypeptide backbone folding in dihydrofolate reductase. The innermost four strands and two bridging helices are geometrically similar to but are connected in a different way from those in the dinucleotide binding domains found in nicotinamide-adenine dinucleotide-linked dehydrogenases. Methotrexate is bound in a 15-angstrom-deep cavity with the pteridine ring buried in a primarily hydrophobic pocket, although a strong interaction occurs between the side chain of aspartic acid 27 and N(1), N(8), and the 2-amino group of methotrexate.
Sigma-Aldrich
Methotrexate hydrate, ≥99.0% (sum of enantiomers, HPLC)