Merck
  • Sorption of benzidine and 3,3'-dichlorobenzidine to lake sediments. 1. Conceptualization and development of a multiparameter model.
Sorption of benzidine and 3,3'-dichlorobenzidine to lake sediments. 1. Conceptualization and development of a multiparameter model.
Environmental toxicology and chemistry (2005-08-23)
Francis P Donaldson, Marianne C Nyman
ABSTRACT
Aromatic amines, such as benzidine and 3,3'-dichlorobenzidine (DCB), are part of the dyes and pigments manufacturing process. The prolonged use of these carcinogenic chemicals in the past generation has introduced a significant amount of contamination to the environment. Their persistency in several mediums has sparked a number of studies in an attempt to develop predictive tools of their fate and transport in the environment. In this study, benzidine and DCB batch isotherms were developed and evaluated. The sediment samples were variable in composition, ranging from sandy to silty-clay sediment samples. The batch isotherms were then analyzed using high-performance liquid chromatography. Subsequently, a multiparameter model (MPM) that accounted for partitioning, covalent bonding, and cation exchange was developed and tested in an effort to understand the various mechanisms. Results proved the proposed model to be effective in predicting sorption of aromatic amines to lake sediments. The findings suggest that the MPM can provide a better understanding of the sorption process of aromatic amines than more conventional models.
Product Description
Supelco
3,3′-Dichlorobenzidine solution, certified reference material, 2000 μg/mL in methanol
Supelco
3,3′-Dichlorobenzidine solution, certified reference material, 5000 μg/mL in methanol
Supelco
3,3′-Dichlorobenzidine, analytical standard