Merck
  • Raman spectral analysis of phenoxyacetic acid and some chloro substituted phenoxyacetic acids.
Raman spectral analysis of phenoxyacetic acid and some chloro substituted phenoxyacetic acids.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2005-07-19)
B Karthikeyan, B Saravanan
ABSTRACT
Raman spectra of phenoxyacetic acid and chlorine substituted phenoxyacetic acids viz., o-chlorophenoxyacetic acid, p-chlorophenoxyacetic acid and 2,6-dichlorophenoxyacetic acid have been analysed with the aid of abinitio theoretical calculations. The effect of chlorine substituents on the Raman spectra of phenoxyacetic acid have also been analysed by considering some important vibrational modes.
Sigma-Aldrich
2,6-Dichlorophenol, 99%
Supelco
2,6-Dichlorophenol, PESTANAL®, analytical standard
Supelco
2,6-Dichlorophenol solution, certified reference material, 5000 μg/mL in methanol