Merck
Biologic markers of exposure to chlorinated solvents.
Clinics in occupational and environmental medicine (2004-08-25)
Javier C Waksman, Scott D Phillips
ABSTRACT
This article presents the current knowledge and clinical applications of the use of biomarkers of exposure to the halogenated solvents 1,1,1 trichloroethane (methylchloroform), trichloroethylene, tetrachloroethylene (perchloroethylene), and 1,1 dichloroethylene (vinylidene chloride). Although some studies have shown that protein and DNA adducts may form with chlorinated hydrocarbons, their application has not been validated sufficiently to justify their use as biologic markers of exposure.
Sigma-Aldrich
1,1-Dichloroethylene, contains 200 ppm MEHQ as inhibitor, 99%
Supelco
1,1-Dichloroethene solution, certified reference material, 1000 μg/mL in methanol
Supelco
1,1-Dichloroethylene, analytical standard