Merck
  • A disubstituted succinamide is a potent sodium channel blocker with efficacy in a rat pain model.
A disubstituted succinamide is a potent sodium channel blocker with efficacy in a rat pain model.
Biochemistry (2004-07-28)
Birgit T Priest, Maria L Garcia, Richard E Middleton, Richard M Brochu, Samantha Clark, Ge Dai, Ivy E Dick, John P Felix, Chou J Liu, Brita S Reiseter, William A Schmalhofer, Pengcheng P Shao, Yui S Tang, Margaret Z Chou, Martin G Kohler, McHardy M Smith, Vivien A Warren, Brande S Williams, Charles J Cohen, William J Martin, Peter T Meinke, William H Parsons, Keith A Wafford, Gregory J Kaczorowski
ABSTRACT
Sodium channel blockers are used clinically to treat a number of neuropathic pain conditions, but more potent and selective agents should improve on the therapeutic index of currently used drugs. In a high-throughput functional assay, a novel sodium channel (Na(V)) blocker, N-[[2'-(aminosulfonyl)biphenyl-4-yl]methyl]-N'-(2,2'-bithien-5-ylmethyl)succinamide (BPBTS), was discovered. BPBTS is 2 orders of magnitude more potent than anticonvulsant and antiarrhythmic sodium channel blockers currently used to treat neuropathic pain. Resembling block by these agents, block of Na(V)1.2, Na(V)1.5, and Na(V)1.7 by BPBTS was found to be voltage- and use-dependent. BPBTS appeared to bind preferentially to open and inactivated states and caused a dose-dependent hyperpolarizing shift in the steady-state availability curves for all sodium channel subtypes tested. The affinity of BPBTS for the resting and inactivated states of Na(V)1.2 was 1.2 and 0.14 microM, respectively. BPBTS blocked Na(V)1.7 and Na(V)1.2 with similar potency, whereas block of Na(V)1.5 was slightly more potent. The slow tetrodotoxin-resistant Na(+) current in small-diameter DRG neurons was also potently blocked by BPBTS. [(3)H]BPBTS bound with high affinity to a single class of sites present in rat brain synaptosomal membranes (K(d) = 6.1 nM), and in membranes derived from HEK cells stably expressing Na(V)1.5 (K(d) = 0.9 nM). BPBTS dose-dependently attenuated nociceptive behavior in the formalin test, a rat model of tonic pain. On the basis of these findings, BPBTS represents a structurally novel and potent sodium channel blocker that may be used as a template for the development of analgesic agents.
Sigma-Aldrich
Succinamide, 98%
Pricing and availability is not currently available.