- Small peptides catalyze highly enantioselective direct aldol reactions of aldehydes with hydroxyacetone: unprecedented regiocontrol in aqueous media.
Small peptides catalyze highly enantioselective direct aldol reactions of aldehydes with hydroxyacetone: unprecedented regiocontrol in aqueous media.
Organic letters (2004-06-18)
Zhuo Tang, Zhi-Hua Yang, Lin-Feng Cun, Liu-Zhu Gong, Ai-Qiao Mi, Yao-Zhong Jiang
PMID15200341
ABSTRACT
[reaction: see text] L-Proline-based small peptides have been developed as efficient catalysts for the asymmetric direct aldol reactions of hydroxyacetone with aldehydes. Chiral 1,4-diols 7, which are disfavored products in similar aldol reactions catalyzed by either aldolases or L-proline, were obtained in high yields and enantioselectivities of up to 96% ee with peptides 3 and 4 in aqueous media.