Merck
  • Two stage biological treatment of a diazo reactive textile dye and the fate of the dye metabolites.
Two stage biological treatment of a diazo reactive textile dye and the fate of the dye metabolites.
Chemosphere (2004-05-04)
Judy A Libra, Maren Borchert, Lothar Vigelahn, Thomas Storm
ABSTRACT
A two stage anaerobic/aerobic bacterial process was used to decolorize and partially mineralize a reactive vinyl sulfone diazo dye C.I. Reactive Black 5 (RB5) in a synthetic wastewater. Since the anchor group of reactive dyes reacts during the dyeing process, the effect the degree of hydrolysis of the vinyl sulfone dye had on decolorization, mineralization and toxicity in each stage was investigated. An overall color removal of approximately 65% was found for both the fully and partially hydrolyzed dye. Partial mineralization of the fully hydrolyzed RB5 was achieved in the two stage rotating disc reactors. While the anchor group metabolite p-aminobenzene-2-hydroxyethylsulfonic acid (p-ABHES) was mineralized, an oxidized form of the center metabolite (1,2-ketimino-7-amino-8-hydroxynaphthalene-3,6-disulfonic acid) remained in the aerobic stage effluent, causing the effluent to be colored although no RB5 was present. Partially hydrolyzed dye in the influent with vinyl forms of the anchor group caused cessation of biogas production and a reduction in decolorization efficiency in the anaerobic stage. No evidence for mineralization of the partially hydrolyzed dye or its metabolites was found. A method for evaluating dye mineralization using lumped parameters is presented.
Sigma-Aldrich
Isethionic acid ammonium salt, 99%
Sigma-Aldrich
Isethionic acid sodium salt, 98%