Merck
  • Novel halo-oxo-allenic fatty ester derivatives from epoxidized methyl santalbate (methyl trans-11-octadecen-9-ynoate).
Novel halo-oxo-allenic fatty ester derivatives from epoxidized methyl santalbate (methyl trans-11-octadecen-9-ynoate).
Chemistry and physics of lipids (2003-09-23)
M S F Lie Ken Jie, M M L Lau, C N W Lam, M S Alam, J O Metzger, U Biermann
ABSTRACT
Methyl santalbate (methyl trans-11-octadecen-9-ynoate) from Sandal wood seed oil, Santalbum alum) was epoxidized to methyl trans-11,12-epoxy-octadec-9-ynoate (1). Treatment of compound 1 with tetrabutylammonium dihydrogentrifluoride, and boron trifluoride etherate gave the corresponding anti- (2a) (57%) and syn- (2b) (35%) fluorohydrin derivatives, respectively. These reactions were regio- and stereoselective in nature. The structures of the anti- and syn- isomers were confirmed by NMR spectroscopy. Ring opening of the epoxy system of compound 1 with lithium chloride gave the anti-chlorohydrin derivative (3) (89%). Oxidation of either compound 2a or 2b gave the same fluoro-keto acetylenic fatty ester (4) (75%), and compound 3 on chromic acid oxidation yielded the corresponding chloro-keto acetylene (5) (73%). Isomerization of compounds 4 and 5 with potassium carbonate in dichloromethane furnished the requisite fluoro-allenic (6) (63%, methyl 11-fluoro-12-oxo-9,10-octadecadienoate) and chloro-allenic (7) (80%, methyl 11-chloro-12-oxo-9,10-octadecadienoate) C(18) fatty esters. All products were confirmed by a combination of spectrometric and spectroscopic techniques.
Sigma-Aldrich
Boron trifluoride diethyl etherate
Sigma-Aldrich
Boron trifluoride diethyl etherate, purified by redistillation, ≥46.5% BF3 basis