Merck
Molecular weight changes in polymer erosion.
Pharmaceutical research (1992-10-01)
A D'Emanuele, J Hill, J A Tamada, A J Domb, R Langer
ABSTRACT
We report a study of the effects of polymer molecular weight on the erosion of polyanhydride copolymer matrices composed of 1,3-bis(p-carboxyphenoxy)-propane (CPP) and sebacic acid (SA) in aqueous solution. The erosion profile characteristically displays an induction period during which the erosion rate is relatively slow. The length of this period depends on the initial molecular weight of the polymer. The induction period may be characterized as a time during which a rapid decrease in polymer molecular weight occurs, the end of this period correlating with the time required for the polymer molecular weight to decrease to below a value of approximately 5000 (MW).
Sigma-Aldrich
1,3-Bis(4-carboxyphenoxy)propane, 97%
Pricing and availability is not currently available.