Merck
  • Predominance of oxidative deamination in the metabolism of exogenous noradrenaline by the normal and chemically denervated human uterine artery.
Predominance of oxidative deamination in the metabolism of exogenous noradrenaline by the normal and chemically denervated human uterine artery.
Naunyn-Schmiedeberg's archives of pharmacology (1992-09-01)
D Branco, M Caramona, F Martel, J A de Almeida, W Osswald
ABSTRACT
Longitudinal strips were prepared from human uterine arteries obtained at hysterectomy. The artery had a low content of noradrenaline and dopamine, contrasting with a high content of the deaminated catechols, dihydroxyphenylglycol (DOPEG) and dihydroxymandelic acid (DOMA), which together represented 98% of endogenous catechols. When incubated with 3H-noradrenaline (0.1 mumol/l), the uterine artery removed, accumulated and metabolized noradrenaline. Deaminated metabolites predominated, DOMA being the most abundant metabolite. Cocaine markedly reduced the accumulation of 3H-noradrenaline and abolished 3H-DOPEG formation, but did not change 3H-DOMA. Selective monoamine oxidase (MAO) inhibitors (clorgyline, selegiline and 2-amino ethyl carboxamide derivatives) caused a marked decrease in the amounts of 3H-DOPEG, 3H-DOMA and 3H-O-methylated and deaminated metabolites (OMDA) formed by the tissue and an increase in 3H-normetanephrine (NMN) formation. Inhibition of catechol-O-methyltransferase suppressed NMN formation and reduced that of OMDA; hydrocortisone slightly depressed the formation of DOMA and OMDA. Homogenates of the uterine artery deaminated 3H-5-HT, 14C-phenylethylamine and 3H-tyramine; inhibition curves of the deamination of 3H-tyramine by clorgyline and selegiline were compatible with the presence of both MOA A and MOA B. Exposure of the strips to 6-hydroxydopamine (1.5 mmol/l for 20 min; 3 exposure periods followed by washout periods of 15,15 and 30 min) resulted in complete and selective chemical denervation of the arterial tissue. This chemical denervation had effects which were similar to those of cocaine. The 2-amino ethyl carboxyamide derivatives markedly reduced the formation of deaminated metabolites by the denervated strips.(ABSTRACT TRUNCATED AT 250 WORDS)
Sigma-Aldrich
DL-3,4-Dihydroxymandelic acid, 95%