Merck
  • [The use of local iodobromine baths in the early sanatorium rehabilitation of myocardial infarct patients with arterial hypertension].
[The use of local iodobromine baths in the early sanatorium rehabilitation of myocardial infarct patients with arterial hypertension].
Likars'ka sprava (1992-01-01)
L E Mikhno, S A Novikov
ABSTRACT
The effect of local iodine-bromine baths at the early stage of health-resort treatment on the main values of the cardio-vascular system was studied in patients suffering of myocardial infarction with arterial hypertension. It was established that local iodine-bromine baths produce a more integrative effects on the cardiovascular system than drugs. They favour the arterial pressure reduction, increase of physical working capacity, normalization of the parameters of central hemodynamics.
Sigma-Aldrich
Iodine monobromide, 98%