Merck
  • [Energy transfer between electric polymer PVK and terbium complex and its transmission electronmicroscope (TME) study].
[Energy transfer between electric polymer PVK and terbium complex and its transmission electronmicroscope (TME) study].
Guang pu xue yu guang pu fen xi = Guang pu (2003-09-10)
D L Tao, B G Huang, Y Z Xu, W H Li, Z L Yang, J G Wu, L D Sun, C H Yan, X X Liu, F X Guo, D F Xu
ABSTRACT
A chloroform-soluble terbium complex, which is confirmed to be Tb(aspirin)3 phen using element analysis and FT-IR spectroscopy, was synthesized. Photoluminescent investigation on the terbium complex and PVK-terbium complex composite was conducted. Förster energy transfer occurred between the terbium complex and the PVK matrix. There are no overlap between UV spectrum of the complex and the emission spectrum of PVK, however, overlap is observed between the excitation spectrum of the complex and the emission spectrum of PVK. Therefore, we suggest that the necessary condition of Förster energy transfer should be overlap between the excitation (not UV) spectrum of one complex and the emission spectrum of polymer matrix. Further investigation indicates that the emission of PVK can be suppressed at different extents by doping various amount of Tb(aspirin)3 phen into PVK films. The ratio of Tb(aspirin)3 phen: PVK = 1:2 (wt%) are regarded as an optimized ratio for limiting the emission of PVK. TEM images of PVK/Tb(aspirin)3 phen films reveal that nanoparticles of the Tb complex are dispersed in the PVK matrix. The size of the aggregated complex in PVK matrix is 20-30 nm. The film is not homogeneous as dark regions co-exist with light region in the TEM images. This phenomenon may be related to the short lifetime of electroluminescent devices.
Sigma-Aldrich
Poly(9-vinylcarbazole), average Mw ~1,100,000, powder
Sigma-Aldrich
Poly(9-vinylcarbazole), average Mn 25,000-50,000