Merck
  • Anti-herbicide single-chain antibody expression confers herbicide tolerance in transgenic plants.
Anti-herbicide single-chain antibody expression confers herbicide tolerance in transgenic plants.
FEBS letters (2003-08-26)
Junichi Eto, Yoshihito Suzuki, Hideo Ohkawa, Isomaro Yamaguchi
ABSTRACT
An anti-chlorpropham single-chain variable-fragment (scFv) gene was introduced into Arabidopsis in a manner to express the antibody fragment in each of four different subcellular compartments. The accumulation of scFv in transgenic plants was detected by targeting the fragment in the endoplasmic reticulum or apoplastic space, or by expressing the fragment as a glycosylphosphatidylinositol-anchored protein, while no accumulation could be detected by targeting the fragment in the cytosol. Transgenic plants accumulating the scFv gene at a high level in the endoplasmic reticulum had enhanced tolerance to chlorpropham in comparison with the non-transformants.
Product Description
Supelco
Chlorpropham, PESTANAL®, analytical standard
Pricing and availability is not currently available.