- Flow-injection chemiluminescence determination of chlorinated isocyanuric acids.
Flow-injection chemiluminescence determination of chlorinated isocyanuric acids.
Analytical and bioanalytical chemistry (2003-02-18)
Afsaneh Safavi, Mohammad Ali Karimi
PMID12589508
ABSTRACT
A rapid and sensitive flow-injection chemiluminescence method is described for the determination of dichloro- and trichloroisocyanuric acids based on the chemiluminescence produced during their reaction with luminol in alkaline medium. The effects of analytical and flow-injection variables on these chemiluminescence systems and determination of both oxidants are discussed. The optimized method yielded 3sigma detection limits of 8x10(-8) and 5x10(-8) mol L(-1) for the sodium dichloroisocyanurate and trichloroisocyanuric acid, respectively. The optimum conditions were found to be as follows: NaOH, 1x10(-1) mol L(-1); luminol, 5x10(-3) mol L(-1); KI, 2x10(-3) mol L(-1) and flow rate, 3.5 mL min(-1).