Merck
  • Pinacol coupling reaction of beta-halo-alpha,beta-unsaturated aldehydes promoted by TiI4.
Pinacol coupling reaction of beta-halo-alpha,beta-unsaturated aldehydes promoted by TiI4.
Organic letters (2002-11-09)
Makoto Shimizu, Hiroshi Goto, Ryuuichirou Hayakawa
ABSTRACT
The pinacol reaction of beta-halogenated alpha,beta-unsaturated aldehydes was promoted by titanium tetraiodide to give coupling products in good yields with high dl-selectivity. Subsequent reduction with H(2)/Pd-C gave saturated vic-diols in good yields. Heck coupling reaction enabled the displacement of halogens with vinyl groups without the loss of stereochemical integrities. [reaction: see text]
Sigma-Aldrich
3,3-Dimethyl-2-butanone, 97%