Merck
  • Teratogenic effects of amitraz, 2,4-dimethylaniline, and paraquat on developing frog (Xenopus) embryos.
Teratogenic effects of amitraz, 2,4-dimethylaniline, and paraquat on developing frog (Xenopus) embryos.
Archives of environmental contamination and toxicology (2002-06-05)
O Osano, A A Oladimeji, M H S Kraak, W Admiraal
ABSTRACT
Developmental effects of amitraz (acaricide), its metabolite (2,4-dimethylaniline), and paraquat (herbicide) on embryos of a nontarget organism, Xenopus laevis, were investigated. Following the standard protocol of the American Society for Testing and Materials (ASTM), the experiments were carried out using native Xenopus frogs. There was a drastic increase in mortality from 24 h to 96 h for paraquat, but 2,4-dimethylaniline showed no mortality at the highest concentration tested (100 mg/L). The 96-h LC(50) values were 0.67, 3.27, and >100 mg/L for paraquat, amitraz, and 2,4-dimethylaniline, respectively. At concentrations higher than 0.2 mg/L of paraquat all the embryos were malformed, whereas growth reduction was apparent at all test concentrations (0.1-5 mg/L). The most common teratogenic effects were flexures of the notochord and stunting of growth. Edema was the most common effect of amitraz on the embryos, and 100% of the surviving embryos in 5 mg/L were edematous. The 96-h EC(50) (malformation) values were 1.21 (95% CI 0.48-3.03) and 0.18 (95% CI 0.16-0.20) mg/L for amitraz and paraquat, respectively. The ratio of 96-h LC(50) to 96-h EC(50) (malformation), i.e., the teratogenicity index (TI) were 2.7 and 3.72 for amitraz and paraquat, respectively, and for 2,4-dimethylaniline (TI > 5) all the embryos in 25 mg/L showed observable pigment loss and encephalomegaly. This shows that paraquat and the degradation product of amitraz, 2,4-dimethylaniline, should be classified as teratogens. Teratogenic risks of massive application of these pesticides on Kenyan farms should therefore be considered.
Sigma-Aldrich
2,4-Dimethylaniline, ≥99%
Supelco
2,4-Dimethylaniline, analytical standard
Sigma-Aldrich
2,4-Dimethylaniline, 98%