Merck
  • Electrospray ionization mass spectrometric studies of phosphorus oxychloride directed synthesis of homo-oligopeptide ester libraries.
Electrospray ionization mass spectrometric studies of phosphorus oxychloride directed synthesis of homo-oligopeptide ester libraries.
Rapid communications in mass spectrometry : RCM (2002-04-23)
Ning Zhou, Kui Lu, Yan Liu, Yi Chen, Guo Tang, Shu-Xia Cao, Ling-Bo Qu, Yu-Fen Zhao
ABSTRACT
With the assistance of phosphorus oxychloride, alpha-amino acids were assembled into homo-peptides, which were analyzed by electrospray ionization mass spectrometry (ESI-MS) and multistage electrospray ionization mass spectrometry (ESI-MS/MS). On quenching with water or various alcohols, the reaction mixtures yielded the corresponding peptides or peptide esters, respectively. This paper reports a simple method to synthesize the homo-oligo-peptide-ester conjugated library by phosphorus oxychloride activation.
Sigma-Aldrich
Phosphorus(V) oxychloride, 99.999%
Sigma-Aldrich
Phosphorus(V) oxychloride, ReagentPlus®, 99%