Merck
  • Oxabicyclo[3.2.1]octenes in organic synthesis: direct ring opening of oxabicyclo[3.2.1] ring systems with diisobutylaluminum hydride and a silyl ketene acetal--synthesis of the chiral C(19)-C(26) and C(27)-C(32) fragments of Scytophycin C.
Oxabicyclo[3.2.1]octenes in organic synthesis: direct ring opening of oxabicyclo[3.2.1] ring systems with diisobutylaluminum hydride and a silyl ketene acetal--synthesis of the chiral C(19)-C(26) and C(27)-C(32) fragments of Scytophycin C.
Organic letters (2002-02-14)
Kevin W Hunt, Paul A Grieco
ABSTRACT
[reaction: see text] An efficient strategy for transforming meso-oxabicyclo[3.2.1]octenone 1 into optically active intermediates for macrolide synthesis has been developed. The direct bridgehead opening of optically active oxabicyclo[3.2.1]octene derivative 2 with hydride or a silyl ketene acetal utilizing the highly polar medium lithium perchlorate in diethyl ether resulted in highly functionalized cycloheptenones, which were transformed into the C(19)-C(26) and C(27)-C(32) fragments of Scytophycin C.
Sigma-Aldrich
(+)-Bis[(R)-1-phenylethyl]amine, 99%
Sigma-Aldrich
Diisobutylaluminum hydride, reagent grade
Sigma-Aldrich
Diisobutylaluminum hydride solution, 1.0 M in hexanes
Sigma-Aldrich
Diisobutylaluminum hydride solution, 1.0 M in cyclohexane
Sigma-Aldrich
Diisobutylaluminum hydride solution, 1.0 M in toluene
Sigma-Aldrich
Diisobutylaluminum hydride solution, 1.0 M in THF
Sigma-Aldrich
Diisobutylaluminum hydride solution, 1.0 M in methylene chloride
Sigma-Aldrich
Diisobutylaluminum hydride solution, 1.0 M in heptane
Sigma-Aldrich
Diisobutylaluminum hydride solution, 25 wt. % in toluene