Merck
  • Defective activity and isoform of the Na,K-ATPase in the dilated cardiomyopathic hamster.
Defective activity and isoform of the Na,K-ATPase in the dilated cardiomyopathic hamster.
Cellular and molecular biology (Noisy-le-Grand, France) (2001-05-17)
F Paganelli, R Mougenot, J M Maixentt
ABSTRACT
The Na,K-ATPase function appears impaired in human heart failure with dilation; however little is known in animal model with idiopathic dilated cardiomyopathy. We studied Na,K-ATPase isoform composition and activity from cardiomyopathic hamsters of the MS 200 strain with pure dilated cardiomyopathy and compared them with those of healthy Syrian hamsters. 150-day-old male MS 200 Syrian hamsters (n = 16) and sex- and age-matched healthy Syrian hamsters (n = 15) were used. Antibodies specific for the three alpha-isoforms and against the beta1-isoform were used to study Na,K-ATPase isoform expression in ventricular myocardium. Na,K-ATPase activity was quantified in homogenate and membrane fractions. There was no significant change in left ventricular mass. Morphological examination revealed a decreased septum thickness in the dilated cardiomyopathy compared with control hamster. Idiopathic dilated cardiomyopathy in hamsters presented significantly reduced membrane alpha1 and beta1 abundances and reduced Na,K-ATPase activity (-35% vs. healthy control, p<0.05). Chronic heart failure had no effect on the Na,K-ATPase alpha2-subunit protein. We have demonstrated for the first time that dilated cardiomyopathy induces a specific reduction of both membrane alpha1- and beta1-isoform abundance and Na,K-ATPase activity in hamsters similar to those previously reported in human dilated heart failure.
Sigma-Aldrich
Digitoxigenin