Merck
  • Red blood cell fatty acid compositions in a patient with autistic spectrum disorder: a characteristic abnormality in neurodevelopmental disorders?
Red blood cell fatty acid compositions in a patient with autistic spectrum disorder: a characteristic abnormality in neurodevelopmental disorders?
Prostaglandins, leukotrienes, and essential fatty acids (2000-09-06)
J G Bell, J R Sargent, D R Tocher, J R Dick
ABSTRACT
The fatty acid compositions of red blood cell (RBC) phospholipids from a patient with autistic spectrum disorder (ASD) had reduced percentages of highly unsaturated fatty acids (HUFA) compared to control samples. The percentage of HUFA in the RBC from the autistic patient was dramatically reduced (up to 70%) when the sample was stored for 6 weeks at -20 degrees C. However, only minor HUFA reductions were recorded in control samples stored similarly, or when the autistic sample was stored at -80 degrees C. A similar instability in RBC HUFA compositions upon storage at -20 degrees C has been recorded in schizophrenic patients. In a number of other neurodevelopmental conditions, including attention deficit hyperactivity disorder (ADHD) and dyslexia, reduced concentrations of RBC HUFA have been recorded. The extent and nature of these aberrations require further assessment to determine a possible common biochemical origin of neurodevelopmental disorders in general. To facilitate this, a large scale assessment of RBC fatty acid compositions in patients with ASD, and related disorders, should be performed as a matter of urgency. Supplementing cells in culture with the tryptophan metabolite indole acrylic acid (IAA) affected the levels of cellular HUFA and prostaglandin production. Indole acroyl glycine (IAG), a metabolite of IAA excreted in urine, is found in high concentrations in patients with neurodevelopmental disorders including ASD, ADHD, dyslexia, Asperger's syndrome and obsessive compulsive disorder.
Sigma-Aldrich
trans-3-Indoleacrylic acid, 98%
Sigma-Aldrich
3-Indoleacrylic acid, BioReagent, ≥98% (HPLC)
Sigma-Aldrich
3-Indoleacrylic acid, powder