Merck
  • Substrate specificity and reaction mechanism of murine 8-oxoguanine-DNA glycosylase.
Substrate specificity and reaction mechanism of murine 8-oxoguanine-DNA glycosylase.
The Journal of biological chemistry (2000-07-08)
D O Zharkov, T A Rosenquist, S E Gerchman, A P Grollman
ABSTRACT
Genomic DNA is prone to oxidation by reactive oxygen species. A major product of DNA oxidation is the miscoding base 8-oxoguanine (8-oxoG). The mutagenic effects of 8-oxoG in mammalian cells are prevented by a DNA repair system consisting of 8-oxoguanine-DNA glycosylase (Ogg1), adenine-DNA glycosylase, and 8-oxo-dGTPase. We have cloned, overexpressed, and characterized mOgg1, the product of the murine ogg1 gene. mOgg1 is a DNA glycosylase/AP lyase belonging to the endonuclease III family of DNA repair enzymes. The AP lyase activity of mOgg1 is significantly lower than its glycosylase activity. mOgg1 releases 8-oxoG from DNA when paired with C, T, or G, but efficient DNA strand nicking is observed only with 8-oxoG:C. Binding of mOgg1 to oligonucleotides containing 8-oxoG:C is strong (K(D) = 51.5 nm), unlike other mispairs. The average residence time for mOgg1 bound to substrate containing 8-oxoG:C is 18.3 min; the time course for accumulation of the NaBH(4)-sensitive intermediate suggests a two-step reaction mechanism. Various analogs of 8-oxoG were tested as substrates for mOgg1. An electron-withdrawing or hydrogen bond acceptor moiety at C8 is required for efficient binding of mOgg1. A substituent at C6 and a keto group at C8 are required for cleavage. The proposed mechanism of 8-oxoG excision involves protonation of O(8) or the deoxyribose oxygen moiety.
Sigma-Aldrich
2′-Deoxyguanosine 5′-triphosphate sodium salt hydrate, ≥96% (HPLC)