Merck
  • Human liver alcohol dehydrogenase: amino acid substitution in the beta 2 beta 2 Oriental isozyme explains functional properties, establishes an active site structure, and parallels mutational exchanges in the yeast enzyme.
Human liver alcohol dehydrogenase: amino acid substitution in the beta 2 beta 2 Oriental isozyme explains functional properties, establishes an active site structure, and parallels mutational exchanges in the yeast enzyme.
Proceedings of the National Academy of Sciences of the United States of America (1984-05-01)
H Jörnvall, J Hempel, B L Vallee, W F Bosron, T K Li
ABSTRACT
The homodimeric Oriental beta 2 beta 2 isozyme of human liver alcohol dehydrogenase, corresponding to an allelic variant at the ADH2 gene locus, was studied in order to define the amino acid exchange in relation to the beta 1 beta 1 isozyme, the predominant allelic form among Caucasians. Sequence analysis reveals that the amino acid substitution occurs at position 7 of the largest CNBr fragment, corresponding to position 47 of the whole protein chain. Here, the beta 2 form has a histidine residue, while, in common with other characterized mammalian liver alcohol dehydrogenases, the beta 1 form has an arginine residue. This exchange does not affect the adjacent cysteine-46 residue, which is a protein ligand to the active-site zinc atom, thus clarifying previously inconsistent results. The histidine/arginine-47 mutational replacement corresponds to a position that binds the pyrophosphate group of the coenzyme NAD(H); this explains the functional differences between the beta 1 beta 1 and beta 2 beta 2 isozymes, including both a lower pH optimum and higher turnover number of beta 2 beta 2, which is likely to be the mutant form. The exchange demonstrates the existence of parallel but separate mutations in the evolution of alcohol dehydrogenases because these mammalian enzymes differ at exactly the same position by the same type of substitution as is found between a mutant and the wild-type constitutive forms of the corresponding yeast enzyme.
Sigma-Aldrich
Alcohol Dehydrogenase equine, recombinant, expressed in E. coli, ≥0.5 U/mg
Sigma-Aldrich
Alcohol Dehydrogenase from Saccharomyces cerevisiae, powder, ≥300 units/mg protein, mol wt ~141,000 (four subunits)
Sigma-Aldrich
Alcohol Dehydrogenase from Saccharomyces cerevisiae
Sigma-Aldrich
Alcohol Dehydrogenase from Saccharomyces cerevisiae, ≥300 units/mg protein, lyophilized powder (contains buffer salts), Mw 141-151 kDa