Merck
  • Human chromosome 11 DNA sequence and analysis including novel gene identification.
Human chromosome 11 DNA sequence and analysis including novel gene identification.
Nature (2006-03-24)
Todd D Taylor, Hideki Noguchi, Yasushi Totoki, Atsushi Toyoda, Yoko Kuroki, Ken Dewar, Christine Lloyd, Takehiko Itoh, Tadayuki Takeda, Dae-Won Kim, Xinwei She, Karen F Barlow, Toby Bloom, Elspeth Bruford, Jean L Chang, Christina A Cuomo, Evan Eichler, Michael G FitzGerald, David B Jaffe, Kurt LaButti, Robert Nicol, Hong-Seog Park, Christopher Seaman, Carrie Sougnez, Xiaoping Yang, Andrew R Zimmer, Michael C Zody, Bruce W Birren, Chad Nusbaum, Asao Fujiyama, Masahira Hattori, Jane Rogers, Eric S Lander, Yoshiyuki Sakaki
ABSTRACT
Chromosome 11, although average in size, is one of the most gene- and disease-rich chromosomes in the human genome. Initial gene annotation indicates an average gene density of 11.6 genes per megabase, including 1,524 protein-coding genes, some of which were identified using novel methods, and 765 pseudogenes. One-quarter of the protein-coding genes shows overlap with other genes. Of the 856 olfactory receptor genes in the human genome, more than 40% are located in 28 single- and multi-gene clusters along this chromosome. Out of the 171 disorders currently attributed to the chromosome, 86 remain for which the underlying molecular basis is not yet known, including several mendelian traits, cancer and susceptibility loci. The high-quality data presented here--nearly 134.5 million base pairs representing 99.8% coverage of the euchromatic sequence--provide scientists with a solid foundation for understanding the genetic basis of these disorders and other biological phenomena.
Sigma-Aldrich
Lactic Dehydrogenase, recombinant from E. coli, ≥90 U/mg
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, 1000 units/mL
Sigma-Aldrich
Phosphatase, Acid from sweet potato, ammonium sulfate suspension, ≥10.0 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine muscle, Type X, ammonium sulfate suspension, ≥600 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, Type III, ammonium sulfate suspension, ≥500 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from porcine heart, ammonium sulfate suspension, ≥200 units/mg protein
Sigma-Aldrich
Phosphorylase a from rabbit muscle, lyophilized powder, 20-30 units/mg protein
Sigma-Aldrich
Phosphatase, Acid from potato, lyophilized powder, ≥3.0 units/mg solid
Sigma-Aldrich
Phosphatase, Acid from potato, lyophilized powder, ≥0.5 unit/mg solid
Sigma-Aldrich
Sphingomyelinase from Staphylococcus aureus, buffered aqueous glycerol solution, 100-300 units/mg protein (Lowry)
Sigma-Aldrich
L-Lactic Dehydrogenase from bovine heart, Type XVII, buffered aqueous glycerol solution, ≥400 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from rabbit muscle, Type II, ammonium sulfate suspension, 800-1,200 units/mg protein
Sigma-Aldrich
L-Lactic Dehydrogenase from rabbit muscle, Type XI, lyophilized powder, 600-1,200 units/mg protein
Sigma-Aldrich
Sphingomyelinase from Bacillus cereus, buffered aqueous glycerol solution, ≥100 units/mg protein (Lowry)
Sigma-Aldrich
Phosphatase, Acid from wheat germ, ≥0.4 unit/mg solid
Sigma-Aldrich
Sphingomyelinase from Bacillus cereus, lyophilized powder, ≥100 units/mg protein
Sigma-Aldrich
Phosphorylase b from rabbit muscle, lyophilized powder, ≥20 units/mg protein, 2× crystallization
Sigma-Aldrich
Phosphorylase b from rabbit muscle, For use as a marker in SDS-PAGE