Merck
  • Determination of haloacetic acids in human urine by headspace gas chromatography-mass spectrometry.
Determination of haloacetic acids in human urine by headspace gas chromatography-mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2010-06-15)
M J Cardador, M Gallego
ABSTRACT
Haloacetic acids (HAAs) are water disinfection byproducts (DBPs) formed by the reaction of chlorine oxidizing compounds with natural organic matter in water containing bromine. HAAs are second to trihalomethanes as the most commonly detected DBPs in surface drinking water and swimming pools. After oral exposure (drinking, showering, bathing and swimming), HAAs are rapidly absorbed from the gastrointestinal tract and excreted in urine. Typical methods used to determine these compounds in urine (mainly from rodents) only deal with one or two HAAs and their sensitivity is inadequate to determine HAA levels in human urine, even those manual sample preparation protocols which are complex, costly, and neither handy nor amenable to automation. In the present communication, we report on a sensitive and straightforward method to determine the nine HAAs in human urine using static headspace (HS) coupled with GC-MS. Important parameters controlling derivatisation and HS extraction were optimised to obtain the highest sensitivity: 120 microl of dimethylsulphate and 100 microl of tetrabutylammonium hydrogen sulphate (derivatisation regents) were selected, along with an excess of Na(2)SO(4) (6 g per 12 ml of urine), an oven temperature of 70 degrees C and an equilibration time of 20 min. The method developed renders an efficient tool for the precise and sensitive determination of the nine HAAs in human urine (RSDs ranging from 6 to 11%, whereas LODs ranged from 0.01 to 0.1 microg/l). The method was applied in the determination of HAAs in urine from swimmers in an indoor swimming pool, as well as in that of non-swimmers. HAAs were not detected in the urine samples from non-swimmers and those of volunteers before their swims; therefore, the concentrations found after exposure were directly related to the swimming activity. The amounts of MCAA, DCAA and TCAA excreted from all swimmers are related to the highest levels in the swimming pool water.
Product Description
Supelco
Dimethyl sulfate, for GC derivatization, LiChropur, ≥99.0% (GC)