Merck
  • The constitutive production of pectinase by the CT1 mutant of Penicillium occitainis is modulated by pH.
The constitutive production of pectinase by the CT1 mutant of Penicillium occitainis is modulated by pH.
Applied biochemistry and biotechnology (2012-11-28)
Zamen Ben Romdhane, Hajer Tounsi, Azza Hadj-Sassi, Noomen Hadj-Taieb, Ali Gargouri
ABSTRACT
The aim of the present study was to investigate pectinases production by CT1 mutant of Penicillium occitanis on glucose based media. Two main groups of pectinases were followed: lyases (pectin and pectate lyases) and hydrolases (polygalacturonases and polymethylgalacturonases). When cultivated in different liquid media, where either the starting glucose concentration or the nature of nitrogen sources used was varied, the CT1 mutant secreted either lyases or hydrolases. In fact, the pH of these various media seemed to correlate with the activity produced: The lyases were highly and exclusively produced at neutral or alkaline ambient pH, whereas hydrolases were highly produced on acidic ambient pH. Such conclusion was confirmed by following pectinase production in the same culture medium (with the same glucose concentration and the same nitrogen source) set at two initial pH of 4 and 7. Altogether, these results suggest that the pectinases control by PacC signaling pathway of P. occitanis should resemble to that of Aspergillus and its ability to "activate the expression of alkaline-expressed genes and repress acid-expressed genes" remains intact in the CT1 over-producing and constitutive strain. Enzymes produced at acidic pH (hydrolases) and at neutral pH (lyases) were applied in the hydrolysis of orange peel and gave results comparable to commercial enzymes.
Sigma-Aldrich
Pectinase from Aspergillus niger, powder, slightly beige, >1 U/mg
Sigma-Aldrich
Pectinase from Aspergillus niger, BioReagent, suitable for plant cell culture, aqueous glycerol solution, ≥5 units/mg protein (Lowry)
Sigma-Aldrich
Pectolyase from Aspergillus japonicus, suitable for plant cell culture, lyophilized powder
Sigma-Aldrich
Pectolyase from Aspergillus japonicus, lyophilized powder, ≥0.3 units/mg solid
Sigma-Aldrich
Pectinase from Rhizopus sp., powder, 400-800 units/g solid