Merck
  • Autoantibodies contribute to the immunopathogenesis of experimental dry eye disease.
Autoantibodies contribute to the immunopathogenesis of experimental dry eye disease.
Investigative ophthalmology & visual science (2012-03-08)
Michael E Stern, Chris S Schaumburg, Karyn F Siemasko, Jianping Gao, Larry A Wheeler, Devin A Grupe, Cintia S De Paiva, Virginia L Calder, Margarita Calonge, Jerry Y Niederkorn, Stephen C Pflugfelder
ABSTRACT
The purpose of this study was to determine if autoantibodies play a role in the immunopathogenesis of experimental dry eye disease. Dry eye was induced by exposing female C57BL/6 wild-type mice or hen egg lysozyme B-cell receptor transgenic mice to desiccating stress (subcutaneous scopolamine [0.5 mg/0.2 mL] 3 times a day, humidity < 40%, and sustained airflow) for 3 weeks, allowing sufficient time for a humoral immune response. Serum or purified IgG isolated from dry-eye mice or untreated controls was passively transferred to nude recipient mice, which were evaluated for ocular surface inflammation 3 days after transfer. To determine if complement activation contributed to serum-induced dry eye disease, cobra venom factor was used to deplete complement activity. Autoantibodies against kallikrein 13 were identified in serum from dry-eye mice, but were undetectable in untreated controls. Autoantibody-containing serum or purified IgG from dry-eye mice was sufficient to mediate complement-dependent ocular surface inflammation. Serum or purified IgG caused marked inflammatory burden and tissue damage within the ocular surface tissues, including elevated Gr1+ neutrophil infiltration and proinflammatory cytokines/chemokines associated with goblet cell loss. Moreover, complement C3b deposition was found within the ocular surface tissues of mice receiving dry-eye serum, but not in recipients of control serum. Functionally, complement depletion attenuated the ability to transfer dry-eye-specific serum or IgG-mediated disease. These data demonstrate for the first time a complement-dependent pathogenic role of dry-eye-specific autoantibodies, and suggest autoantibody deposition within the ocular surface tissues contributes to the predominantly T-cell-mediated immunopathogenesis of dry eye disease.
Sigma-Aldrich
Kallikrein from porcine pancreas, ≥40 units/mg protein
Sigma-Aldrich
Endoproteinase Arg-C from mouse submaxillary gland, suitable for protein sequencing, lyophilized powder