Merck
DREAM mediated regulation of GCM1 in the human placental trophoblast.
PloS one (2013-01-10)
Dora Baczyk, Mark Kibschull, Britt Mellstrom, Khrystyna Levytska, Marcos Rivas, Sascha Drewlo, Stephen J Lye, Jose R Naranjo, John C P Kingdom
ABSTRACT
The trophoblast transcription factor glial cell missing-1 (GCM1) regulates differentiation of placental cytotrophoblasts into the syncytiotrophoblast layer in contact with maternal blood. Reduced placental expression of GCM1 and abnormal syncytiotrophoblast structure are features of hypertensive disorder of pregnancy--preeclampsia. In-silico techniques identified the calcium-regulated transcriptional repressor--DREAM (Downstream Regulatory Element Antagonist Modulator)--as a candidate for GCM1 gene expression. Our objective was to determine if DREAM represses GCM1 regulated syncytiotrophoblast formation. EMSA and ChIP assays revealed a direct interaction between DREAM and the GCM1 promoter. siRNA-mediated DREAM silencing in cell culture and placental explant models significantly up-regulated GCM1 expression and reduced cytotrophoblast proliferation. DREAM calcium dependency was verified using ionomycin. Furthermore, the increased DREAM protein expression in preeclamptic placental villi was predominantly nuclear, coinciding with an overall increase in sumolylated DREAM and correlating inversely with GCM1 levels. In conclusion, our data reveal a calcium-regulated pathway whereby GCM1-directed villous trophoblast differentiation is repressed by DREAM. This pathway may be relevant to disease prevention via calcium-supplementation.
Sigma-Aldrich
LuminoCt® SYBR® Green qPCR ReadyMix, For fast SYBR Green quantitative PCR
Sigma-Aldrich
LuminoCt® qPCR ReadyMix, For fast probe-based quantitative PCR