Merck
  • Low potential detection of glutamate based on the electrocatalytic oxidation of NADH at thionine/single-walled carbon nanotubes composite modified electrode.
Low potential detection of glutamate based on the electrocatalytic oxidation of NADH at thionine/single-walled carbon nanotubes composite modified electrode.
Biosensors & bioelectronics (2008-10-24)
Ling Meng, Ping Wu, Guoxiang Chen, Chenxin Cai, Yongming Sun, Zhenhong Yuan
ABSTRACT
A glutamate biosensor based on the electrocatalytic oxidation of reduced nicotinamide adenine dinucleotide (NADH), which was generated by the enzymatic reaction, was developed via employing a single-walled carbon nanotubes/thionine (Th-SWNTs) nanocomposite as a mediator and an enzyme immobilization matrix. The biosensor, which was fabricated by immobilizing glutamate dehydrogenase (GlDH) on the surface of Th-SWNTs, exhibited a rapid response (ca. 5s), a low detection limit (0.1 microM), a wide and useful linear range (0.5-400 microM), high sensitivity (137.3+/-15.7) microA mM(-1)cm(-2), higher biological affinity, as well as good stability and repeatability. In addition, the common interfering species, such as ascorbic acid, uric acid, and 4-acetamidophenol, did not cause any interference due to the use of a low operating potential (190 mV vs. NHE). The biosensor can be used to quantify the concentration of glutamate in the physiological level. The Th-SWNTs system represents a simple and effective approach to the integration of dehydrogenase and electrodes, which can provide analytical access to a large group of enzymes for wide range of bioelectrochemical applications including biosensors and biofuel cells.
Sigma-Aldrich
Thionin acetate salt, for microscopy (Bact., Bot., Hist.)