Merck
  • pH-independent release of a basic drug from pellets coated with the extended release polymer dispersion Kollicoat SR 30 D and the enteric polymer dispersion Kollicoat MAE 30 DP.
pH-independent release of a basic drug from pellets coated with the extended release polymer dispersion Kollicoat SR 30 D and the enteric polymer dispersion Kollicoat MAE 30 DP.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2004-06-23)
A Dashevsky, K Kolter, R Bodmeier
ABSTRACT
The objective of this study was to obtain pH-independent release profiles from coated pellets containing drugs with pH-dependent solubility. pH-independent release of the basic model drug verapamil HCl was achieved by coating with a combination of the neutral polymer dispersions Kollicoat SR 30 D (aqueous dispersion of polyvinyl acetate) and the enteric polymer dispersion Kollicoat MAE 30 DP (aqueous dispersion of methacrylic acid and ethyl acrylate copolymer; methacrylic acid copolymer type C). The two polymers where applied either as separate layers (enteric polymer + extended release polymer or vice versa) or as a polymer blend. A careful balance of the ratios of the polymers allowed the achievement of a pH-independent release. Higher amounts of the enteric polymer in the polymer blend resulted in a reversal of the pH-dependency, e.g. a faster release at pH 6.8 than in 0.1 N HCl.
Sigma-Aldrich
Kollicoat® MAE 30 DP, 46.0-50.6% methacrylic acid basis (calc. on solids content)
Pricing and availability is not currently available.