Merck
  • Presynaptic GABAB receptors modulate IPSPs evoked in neurons of deep cerebellar nuclei in vitro.
Presynaptic GABAB receptors modulate IPSPs evoked in neurons of deep cerebellar nuclei in vitro.
Journal of neurophysiology (1996-02-01)
D Mouginot, B H Gähwiler
ABSTRACT
1. Recording from deep cerebellar nuclei neurons, we investigated the role of presynaptic gamma-aminobutyric acid-B (GABAB) receptors in the modulation of monosynaptic inhibitory postsynaptic potentials (IPSPs) evoked by stimulation of Purkinje cells in rat slice cultures. 2. Bath application of the GABAB receptor agonist, baclofen (10 and 100 microM) induced two effects in cerebellar nuclei neurons: a postsynaptic hyperpolarization of 4.2 +/- 1.7 (SD) mV and a reduction in the amplitude of evoked IPSPs (30 +/- 10%). 3. When the postsynaptic GABAB response was blocked by filling the electrode with cesium methanesulfonate (2 M), or with a solution containing QX 314 (50 mM), bath application of baclofen (10 microM) reversibly depressed the evoked IPSPs by 36.7 +/- 18.7% and 42 +/- 20.3%, respectively. Under these experimental conditions, baclofen (10 microM) also reduced the amplitude of spontaneous IPSPs (10.2 +/- 9.5%) and decreased their frequency by 45.6 +/- 8.8%, suggesting a presynaptic site of action. 4. The presynaptic action of baclofen was not due to activation of receptors on the somata of Purkinje cells: baclofen (100 microM) failed to alter membrane holding current in Purkinje cells, and it had no effect on the rate of spontaneous action-potential discharge in Purkinje cells in the presence of ionotropic glutamate receptor antagonists (6-cyano-7-nitroquinoxaline-2,3-dione, 20 microM; D-2-amino-5-phosphonovalerate, 40 microM). 5. IPSPs could be evoked by extracellular stimulation of the Purkinje cell layer or by direct stimulation of the fiber bundle connecting Purkinje cells to deep cerebellar neurons. In both situations, baclofen (10 microM) reduced the amplitude of evoked IPSPs by 32.7 +/- 8.8% and 31.2 +/- 10.2%, respectively. 6. These results demonstrate that GABAB receptors are present on the terminals of Purkinje cells. Their activation causes a decrease in the amplitude of evoked IPSPs recorded in deep cerebellar nuclei and also reduces the frequency of spontaneous inhibitory events.
Sigma-Aldrich
Cesium methanesulfonate, used in patch clamp techniques
Pricing and availability is not currently available.