Merck
  • Dapagliflozin protects against chronic heart failure in mice by inhibiting macrophage-mediated inflammation, independent of SGLT2.
Dapagliflozin protects against chronic heart failure in mice by inhibiting macrophage-mediated inflammation, independent of SGLT2.
Cell reports. Medicine (2023-12-21)
Qingqing Wu, Qi Yao, Tongtong Hu, Jiabin Yu, Kebing Jiang, Ying Wan, Qizhu Tang
ABSTRACT
The specific mechanism of sodium-glucose cotransporter 2 (SGLT2) inhibitor in heart failure (HF) needs to be elucidated. In this study, we use SGLT2-global-knockout (KO) mice to assess the mechanism of SGLT2 inhibitor on HF. Dapagliflozin ameliorates both myocardial infarction (MI)- and transverse aortic constriction (TAC)-induced HF. Global SGLT2 deficiency does not exert protection against adverse remodeling in both MI- and TAC-induced HF models. Dapagliflozin blurs MI- and TAC-induced HF phenotypes in SGLT2-KO mice. Dapagliflozin causes major changes in cardiac fibrosis and inflammation. Based on single-cell RNA sequencing, dapagliflozin causes significant differences in the gene expression profile of macrophages and fibroblasts. Moreover, dapagliflozin directly inhibits macrophage inflammation, thereby suppressing cardiac fibroblasts activation. The cardio-protection of dapagliflozin is blurred in mice treated with a C-C chemokine receptor type 2 antagonist. Taken together, the protective effects of dapagliflozin against HF are independent of SGLT2, and macrophage inhibition is the main target of dapagliflozin against HF.
Sigma-Aldrich
CCR2 Antagonist, The CCR2 Antagonist, also referenced under CAS 445479-97-0, controls the biological activity of CCR2.
Pricing and availability is not currently available.