Merck
  • Derivation and transcriptional reprogramming of border-forming wound repair astrocytes after spinal cord injury or stroke in mice.
Derivation and transcriptional reprogramming of border-forming wound repair astrocytes after spinal cord injury or stroke in mice.
Nature neuroscience (2024-06-22)
Timothy M O'Shea, Yan Ao, Shinong Wang, Yilong Ren, Amy L Cheng, Riki Kawaguchi, Zechuan Shi, Vivek Swarup, Michael V Sofroniew
ABSTRACT
Central nervous system (CNS) lesions become surrounded by neuroprotective borders of newly proliferated reactive astrocytes; however, fundamental features of these cells are poorly understood. Here we show that following spinal cord injury or stroke, 90% and 10% of border-forming astrocytes derive, respectively, from proliferating local astrocytes and oligodendrocyte progenitor cells in adult mice of both sexes. Temporal transcriptome analysis, single-nucleus RNA sequencing and immunohistochemistry show that after focal CNS injury, local mature astrocytes dedifferentiate, proliferate and become transcriptionally reprogrammed to permanently altered new states, with persisting downregulation of molecules associated with astrocyte-neuron interactions and upregulation of molecules associated with wound healing, microbial defense and interactions with stromal and immune cells. These wound repair astrocytes share morphologic and transcriptional features with perimeningeal limitans astrocytes and are the predominant source of neuroprotective borders that re-establish CNS integrity around lesions by separating neural parenchyma from stromal and immune cells as occurs throughout the healthy CNS.
Sigma-Aldrich
Anti-HA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Oligodendrocyte transcription factor 2 (OLIG2 Antibody), serum, from guinea pig