Merck
  • Skewed X-inactivation is associated with retinal dystrophy in female carriers of RPGR mutations.
Skewed X-inactivation is associated with retinal dystrophy in female carriers of RPGR mutations.
Life science alliance (2023-08-05)
Muhammad Usman, Christoph Jüschke, Fei Song, Dennis Kastrati, Marta Owczarek-Lipska, Jannis Eilers, Laurenz Pauleikhoff, Clemens Lange, John Neidhardt
ABSTRACT
Progressive degeneration of rod and cone photoreceptors frequently is caused by mutations in the X-chromosomal gene Retinitis Pigmentosa GTPase Regulator (RPGR). Males hemizygous for a RPGR mutation often are affected by Retinitis Pigmentosa (RP), whereas female mutation carriers only occasionally present with severe RP phenotypes. The underlying pathomechanism leading to RP in female carriers is not well understood. Here, we analyzed a three-generation family in which two of three female carriers of a nonsense RPGR mutation presented with RP. Among two cell lines derived from the same female family members, differences were detected in RPGR transcript expression, in localization of RPGR along cilia, as well as in primary cilium length. Significantly, these differences correlated with alterations in X-chromosomal inactivation patterns found in the patient-derived cell lines from females. In summary, our data suggest that skewed X-chromosomal inactivation is an important factor that determines the disease manifestation of RP among female carriers of pathogenic sequence alterations in the RPGR gene.
Sigma-Aldrich
Anti-Tubulin Antibody, Detyrosinated, Chemicon®, from rabbit
Sigma-Aldrich
Anti-RPGR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution