Merck
  • Muscarinic Receptor Stimulation Does Not Inhibit Voltage-dependent Ca2+ Channels in Rat Adrenal Medullary Chromaffin Cells.
Muscarinic Receptor Stimulation Does Not Inhibit Voltage-dependent Ca2+ Channels in Rat Adrenal Medullary Chromaffin Cells.
Acta histochemica et cytochemica (2023-09-08)
Keita Harada, Masumi Inoue
ABSTRACT
Adrenal medullary chromaffin (AMC) and sympathetic ganglion cells are derived from the neural crest and show a similar developmental path. Thus, these two cell types have many common properties in membrane excitability and signaling. However, AMC cells function as endocrine cells while sympathetic ganglion cells are neurons. In rat sympathetic ganglion cells, muscarinic M1 and M4 receptors mediate excitation and inhibition via suppression of M-type K+ channels and suppression of voltage-dependent Ca2+ channels, respectively. On the other hand, M1 receptor stimulation in rat AMC cells also produces excitation by suppressing TWIK-related acid sensitive K+ (TASK) channels. However, whether M4 receptors are coupled with voltage-dependent Ca2+ channel suppression is unclear. We explore this issue electrophysiologically and biochemically. Electrical stimulation of nerve fibers in rat adrenal glands trans-synaptically increased the Ca2+ signal in AMC cells. This electrically evoked increased Ca2+ signal was not altered during muscarine-induced increase in Ca2+ signal, whereas it decreased significantly during a GABA-induced increase, due to a shunt effect of increased Cl- conductance. The whole-cell current recordings revealed that voltage-dependent Ca2+ currents in AMC cells were suppressed by adenosine triphosphate, but not by muscarinic agonists. The fractionation analysis and immunocytochemistry indicated that CaV1.2 Ca2+ channels and M4 receptors are located in the raft and non-raft membrane domains, respectively. We concluded that muscarinic stimulation in rat AMC cells does not produce voltage-dependent Ca2+ channel inhibition. This lack of muscarinic inhibition is at least partly due to physical separation of voltage-dependent Ca2+ channels and M4 receptors in the plasma membrane.
Sigma-Aldrich
Anti-Muscarinic Acetylcholine Receptor m4 Antibody, clone 17F10.2, purified antibody, clone 17F10.2, Chemicon®