Merck
  • Potential roles of neurotensin on cognition in conditions of obese-insulin resistance.
Potential roles of neurotensin on cognition in conditions of obese-insulin resistance.
Neuropeptides (2018-10-04)
Napatsorn Saiyasit, Jirapas Sripetchwandee, Nipon Chattipakorn, Siriporn C Chattipakorn
ABSTRACT
Neurotensin is an endogenous tridecapeptide that can be found in both central and peripheral nervous systems. Under normal physiological conditions, neurotensin is involved in the regulation of pain, body temperature, physical activity, appetite as well as learning and memory. In addition, it plays an important role in fat metabolism. Previous studies have demonstrated that alterations of neurotensin levels were associated with several neuropathological conditions such as Alzheimer's disease, mood disorders, and obesity associated eating disorders. Obesity has been shown to be associated with low-grade systemic inflammation, brain inflammation, and cognitive decline. Several pieces of evidence suggest that neurotensin might play a role in cognitive decline following obesity. However, the underlying mechanisms of neurotensin on cognition under obese-insulin resistant condition are still unclear. In this review, the current available evidence from in vitro, in vivo and clinical studies regarding the role of neurotensin in the physiological condition and obesity in association with cognition are comprehensively summarized and discussed. The studies which report controversial findings regarding these issues are also presented and discussed.
Sigma-Aldrich
Neurotensin, Potent hypothalamic peptide neurotransmitter.
Pricing and availability is not currently available.