Merck
  • Standalone cell culture microfluidic device-based microphysiological system for automated cell observation and application in nephrotoxicity tests.
Standalone cell culture microfluidic device-based microphysiological system for automated cell observation and application in nephrotoxicity tests.
Lab on a chip (2023-12-22)
Hiroshi Kimura, Hiroko Nakamura, Tomomi Goto, Wakana Uchida, Takayuki Uozumi, Daniel Nishizawa, Kenta Shinha, Junko Sakagami, Kotaro Doi
ABSTRACT
Microphysiological systems (MPS) offer an alternative method for culturing cells on microfluidic platforms to model organ functions in pharmaceutical and medical sciences. Although MPS hardware has been proposed to maintain physiological organ function through perfusion culture, no existing MPS can automatically assess cell morphology and conditions online to observe cellular dynamics in detail. Thus, with this study, we aimed to establish a practical strategy for automating cell observation and improving cell evaluation functions with low temporal resolution and throughput in MPS experiments. We developed a versatile standalone cell culture microfluidic device (SCCMD) that integrates microfluidic chips and their peripherals. This device is compliant with the ANSI/SLAS standards and has been seamlessly integrated into an existing automatic cell imaging system. This integration enables automatic cell observation with high temporal resolution in MPS experiments. Perfusion culture of human kidney proximal tubule epithelial cells using the SCCMD improves cell function. By combining the proximal tubule MPS with an existing cell imaging system, nephrotoxicity studies were successfully performed to automate morphological and material permeability evaluation. We believe that the concept of building the ANSI/SLAS-compliant-sized MPS device proposed herein and integrating it into an existing automatic cell imaging system for the online measurement of detailed cell dynamics information and improvement of throughput by automating observation operations is a novel potential research direction for MPS research.
Sigma-Aldrich
Anti-SLC47A1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Pricing and availability is not currently available.