Merck
  • Vertical mother-to-infant transmission of herpes simplex virus 2 is correlated with tropism due to mutations in viral UL13.
Vertical mother-to-infant transmission of herpes simplex virus 2 is correlated with tropism due to mutations in viral UL13.
Journal of medical virology (2024-01-18)
Kimiyasu Shiraki, Tohru Daikoku, Mark N Prichard, Koma Matsuo, Tomoko Okuda, Yoshihiro Yoshida, Masaya Takemoto, Kenji Takeuchi, Kiyonao Sada, Richard Whitley, Takashi Kawana
ABSTRACT
Although neonates are commonly exposed to vaginal herpes simplex virus (HSV)-2, neonatal herpes is rare. Therefore, we analyzed paired infant and maternal HSV-2 isolates from two cases of mother-to-infant transmission to identify viral factors contributing to vertical transmission. Sixteen infant isolates with neonatal herpes and 27 genital isolates in their third trimester were included. The infant isolates were significantly more temperature-independent than the maternal isolates. Sequence comparison revealed viral UL13 protein kinase (UL13-PK) mutation in the infant isolates in both cases. In the expanded cohort, infant isolates (5/18) had significantly more UL13-PK mutations than genital isolates (1/29). Isolates within 8 days post-birth (3/4) had a significantly higher frequency of UL13-PK mutation than those after 9 days (2/14), suggesting a close association between UL13-PK mutations and vertical transmission. Elongation factor 1-delta was identified as a target of UL13-PK by proteomic analysis of UL13-PK-positive and -negative HepG2 cells. The mixed infant isolates with the intact and mutated UL13-PK conferred altered cell tropism, temperature independence adapting to fetal temperature, and better growth properties in Vero and hepatoblastoma HepG2 cells than in HSV-2 with intact and mutated UL13-PK alone, indicating that viral UL13-PK mutation is essential for vertical HSV-2 transmission.
Sigma-Aldrich
Anti-Tubulin β antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Tropomyosin 4 Antibody, serum, Chemicon®