Merck
  • Downregulation of Exosomal hsa-miR-551b-3p in Obesity and Its Link to Type 2 Diabetes Mellitus.
Downregulation of Exosomal hsa-miR-551b-3p in Obesity and Its Link to Type 2 Diabetes Mellitus.
Non-coding RNA (2023-11-21)
Kseniia V Dracheva, Irina A Pobozheva, Kristina A Anisimova, Stanislav G Balandov, Maria N Grunina, Zarina M Hamid, Dmitriy I Vasilevsky, Sofya N Pchelina, Valentina V Miroshnikova
ABSTRACT
Obesity is a significant risk factor for the development of type 2 diabetes mellitus (T2DM). Adipose tissue dysfunction can affect the pool of circulating exosomal miRNAs, driving concomitant disease in obesity. These exosomal miRNAs can reflect adipose tissue functionality, thus serving as prognostic biomarkers for disease monitoring in case of T2DM. In the present study, we conducted NanoString microRNA profiling of extracellular vesicles (EVs) secreted by adipose tissue of obese patients (body mass index (BMI) > 35) without T2DM and nonobese individuals (BMI < 30) as a control group. Functional and pathway enrichment analysis showed that miRNAs associated with obesity in this study were implicated in insulin signaling and insulin resistance biological pathways. Further, these microRNAs were screened in serum EVs in the following groups: (1) obese patients with T2DM, (2) obese patients without T2DM, and (3) nonobese individuals as a control group. has-miR-551b-3p was shown to be downregulated in adipose tissue EVs, as well as in serum EVs, of patients with obesity without T2DM. At the same time, the serum exosomal hsa-miR-551b-3p content was significantly higher in obese patients with T2DM when compared with obese patients without T2DM and may be a potential biomarker of T2DM development in obesity.
Sigma-Aldrich
Anti-Omentin Antibody, Chemicon®, from rabbit